Learn More
Biomatik Corporation Integrin β3 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50318
Encompass_Preferred
Integrin β3 Colorimetric Cell-Based ELISA Kit for detecting Integrin β3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05106. Gene ID: 3690. Target Synonyms: CD61; GP3A; GPIIIa; integrin beta-3; ITB3; platelet membrane glycoprotein IIIa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-7023