Learn More
Biomatik Corporation Human N-myc proto-oncogene protein (MYCN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34834
Encompass_Preferred
The Human N-myc proto-oncogene protein (MYCN) ELISA Kit allows for quantitative determination of v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04198. Restriction: For Research Use Only.
Catalog No. 50-149-8122