Learn More
Biomatik Corporation Human Pulmonary surfactant-associated protein B, SP-B ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35299
Encompass_Preferred
The Human Pulmonary surfactant-associated protein B, SP-B ELISA Kit allows for quantitative determination of surfactant protein B in serum, plasma, urine and cell culture supernates. Species Reactivity: Human. Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07988. Restriction: For Research Use Only.
Catalog No. 50-149-8221