Learn More
Biomatik Corporation Human ribonuclease, RNase A family, 2 (liver, eosinophil-derived neurotoxin) (RNASE2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35407
Encompass_Preferred
Human Ribonuclease, Rnase A Family, 2 (Liver, Eosinophil-Derived Neurotoxin) (Rnase2) Elisa Kit for detecting Ribonuclease, Rnase A Family, 2 (Liver, Eosinophil-Derived Neurotoxin) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10153; RNASE2. Target Synonyms: RNASE2; EDN; RNS2; Non-secretory ribonuclease; EC 4.6.1.18; Eosinophil-derived neurotoxin; RNase UpI-2; Ribonuclease 2; RNase 2; Ribonuclease US. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8238