Learn More
Biomatik Corporation Human RING finger protein 207 (RNF207) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35414
Encompass_Preferred
Human Ring Finger Protein 207 (Rnf207) Elisa Kit for detecting Ring Finger Protein 207 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6ZRF8; RNF207. Target Synonyms: RNF207; C1orf188; RING finger protein 207. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8240