Learn More
Biomatik Corporation Human R-spondin-2 (RSPO2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35423
Encompass_Preferred
Human R-Spondin-2 (Rspo2) Elisa Kit for detecting R-Spondin 2 Homolog (Xenopus Laevis) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6UXX9; RSPO2. Target Synonyms: RSPO2; UNQ9384/PRO34209; R-spondin-2; Roof plate-specific spondin-2; hRspo2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8242