Learn More
Biomatik Corporation Human S100 calcium binding protein A9/calgranulin B, S100A9 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35433
Encompass_Preferred
Human S100 Calcium Binding Protein A9/Calgranulin B, S100A9 Elisa Kit for detecting S100 Calcium Binding Protein A9 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4.69-300ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06702; S100A9. Target Synonyms: Leukocyte L1 complex heavy chain; 60B8AG; CAGB; Calgranulin B; Calgranulin-B; Calprotectin L1H subunit; CFAG; CGLB; Cystic fibrosis antigen B; L1AG; Leukocyte L1 complex heavy chain; LIAG; MAC387; MIF; Migration inhibitory factor related protein 14; Migration inhibitory factor-related protein 14; MRP 14; MRP-14; MRP14; Myeloid-related protein 14; NIF; OTTHUMP00000015331; p14; Protein S100-A9; S100 A9; S100 calcium binding protein A9; S100 calcium binding protein A9 calgranulin B; S100 calcium-binding protein A9; S100A9; S10A9_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8244