Learn More
Biomatik Corporation Human Sclerostin (SOST) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35458
Encompass_Preferred
Human Sclerostin (Sost) Elisa Kit for detecting Sclerosteosis in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9BQB4; SOST. Target Synonyms: BEER; CDD; Cortical hyperostosis with syndactyly; Sclerosteosis; Sclerostin; Sost; SOST_HUMAN; SOST1; UNQ2976/PRO7455/PRO7476; VBCH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8249