Learn More
Biomatik Corporation Human Secreted Frizzled-Related Protein 1 (SFRP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35461
Encompass_Preferred
Human Secreted Frizzled-Related Protein 1 (Sfrp1) Elisa Kit for detecting Secreted Frizzled-Related Protein 1 in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8N474; SFRP1. Target Synonyms: Frizzled related protein 1; FRP 1; FRP; FRP-1; FRP1; FrzA; SARP 2; SARP-2; SARP2; Secreted apoptosis related protein 2; Secreted apoptosis-related protein 2; Secreted frizzled related protein 1; Secreted frizzled related protein; Secreted frizzled-related protein 1; SFRP 1; sFRP-1; SFRP1; SFRP1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8250