Learn More
Biomatik Corporation Human secreted frizzled-related protein 5 (SFRP5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35465
Encompass_Preferred
Human Secreted Frizzled-Related Protein 5 (Sfrp5) Elisa Kit for detecting Secreted Frizzled-Related Protein 5 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5T4F7; SFRP5. Target Synonyms: SFRP5; FRP1B; SARP3; Secreted frizzled-related protein 5; sFRP-5; Frizzled-related protein 1b; FRP-1b; Secreted apoptosis-related protein 3; SARP-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8252