Learn More
Biomatik Corporation Human Secretoglobin family 3A member 2 (SCGB3A2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35474
Encompass_Preferred
Human Secretoglobin Family 3A Member 2 (Scgb3A2) Elisa Kit for detecting Secretoglobin, Family 3A, Member 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 46.88-3000pg/ml. Sensitivity: 11.72pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q96PL1; SCGB3A2. Target Synonyms: LU103; Pneumo secretory protein 1; PnSP-1; PNSP1; SCGB3A2; Secretoglobin family 3A member 2; SG3A2_HUMAN; UGRP1; Uteroglobin-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8253