Learn More
Biomatik Corporation Human Secretogranin-2 (SCG2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35476
Encompass_Preferred
Human Secretogranin-2 (Scg2) Elisa Kit for detecting Secretogranin-2(Scg2) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 2.34-150ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13521; SCG2. Target Synonyms: CHGC; Chromogranin C; Chromogranin-C; EM66; SCG2; SCG2_HUMAN; Secretogranin II; Secretogranin-2; Secretoneurin; SGII; SN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8254