Learn More
Biomatik Corporation Human secretory component, SC ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35479
Encompass_Preferred
Human Secretory Component, Sc Elisa Kit for detecting Secretory Component, Sc in serum, plasma, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ug/ml. Sensitivity: 0.078ug/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8255