Learn More
Biomatik Corporation Human Selenoprotein P (SEPP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35484
Encompass_Preferred
Human Selenoprotein P (Sepp1) Elisa Kit for detecting Selenoprotein P, Plasma, 1 in serum, plasma, cell culture supernates, tissue homogenates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49908; SELENOP. Target Synonyms: Selenoprotein P; Selenoprotein P plasma 1; Selp; SeP; Sepp1; SEPP1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8257