Learn More
Biomatik Corporation Human Semaphorin-3D (SEMA3D) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35491
Encompass_Preferred
Human Semaphorin-3D (Sema3D) Elisa Kit for detecting Sema Domain, Immunoglobulin Domain (Ig), Short Basic Domain, Secreted, (Semaphorin) 3D in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95025; SEMA3D. Target Synonyms: Coll 2; Coll2; Collapsin 2; SEM3D_HUMAN; Sema domain immunoglobulin domain (Ig) short basic domain secreted (semaphorin) 3D; Sema Z2; Sema3d; Semaphorin-3D; Semaphorin3D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8258