Learn More
Biomatik Corporation Human Semaphorin-4A (SEMA4A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35494
Encompass_Preferred
Human Semaphorin-4A (Sema4A) Elisa Kit for detecting Sema Domain, Immunoglobulin Domain (Ig), Transmembrane Domain (Tm) And Short Cytoplasmic Domain, (Semaphorin) 4A in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H3S1; SEMA4A. Target Synonyms: CORD10; RP11 54H19 2; RP35; SEM4A_HUMAN; Sema B; Sema domain immunoglobulin domain Ig transmembrane domain TM and short cytoplasmic domain 4A; Sema domain immunoglobulin domain Ig transmembrane domain TM and short cytoplasmic domain semaphorin 4A; Sema domain; immunoglobulin domain (Ig); transmembrane domain (TM) and short cytoplasmic domain; (semaphorin); Sema4a; SEMAB; Semaphorin 4A precursor; Semaphorin B; Semaphorin-4A; Semaphorin-B; SEMB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8259