Learn More
Biomatik Corporation Human Serine protease inhibitor Kazal-type 5 (SPINK5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35505
Encompass_Preferred
Human Serine Protease Inhibitor Kazal-Type 5 (Spink5) Elisa Kit for detecting Serine Peptidase Inhibitor, Kazal Type 5 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NQ38; SPINK5. Target Synonyms: Hemofiltrate peptide HF7665; ISK5_HUMAN; LEKTI; Lympho-epithelial Kazal-type-related inhibitor; Lymphoepithelial Kazal type related inhibitor; NETS; NS; Serine peptidase inhibitor Kazal type 5; SPINK5; VAKTI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8263