Learn More
Biomatik Corporation Human Serine/threonine-protein kinase B-raf (BRAF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35511
Encompass_Preferred
Human Serine/Threonine-Protein Kinase B-Raf (Braf) Elisa Kit for detecting V-Raf Murine Sarcoma Viral Oncogene Homolog B1 in serum, plasma, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P15056; BRAF. Target Synonyms: FLJ95109; 94 kDa B raf protein; B raf 1; B raf; B Raf proto oncogene serine threonine protein kinase; B Raf proto oncogene; serine/threonine kinase; B RAF1; B-Raf proto-oncogene serine/threonine-protein kinase (p94); BRAF 1; BRAF; BRAF_HUMAN; BRAF1; cRmil; MGC126806; MGC138284; Murine sarcoma viral (v-raf) oncogene homolog B1; Murine sarcoma viral v raf oncogene homolog B1; NS7; Oncogen BRAF; oncogene BRAF1; p94; Proto-oncogene B-Raf; Proto-oncogene c-Rmil; RAFB 1; RAFB1; RMIL; Serine/threonine-protein kinase B-raf. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8264