Learn More
Biomatik Corporation Human Serum amyloid A-4 protein (SAA4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35541
Encompass_Preferred
Human Serum Amyloid A-4 Protein (Saa4) Elisa Kit for detecting Serum Amyloid A-4 Protein(Saa4) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35542; SAA4. Target Synonyms: Amyloid A; serum; 4; C SAA; C-SAA; Constitutively expressed serum amyloid A protein; CSAA; SAA 4; Saa4; SAA4 protein; SAA4_HUMAN; Serum amyloid A 4 protein; Serum amyloid A-4 protein; Serum amyloid A4 constitutive. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8266