Learn More
Biomatik Corporation Human Sialidase ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35556
Encompass_Preferred
Human Sialidase Elisa Kit for detecting Sialidase 1 (Lysosomal Sialidase) in serum, urine, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99519; NEU1. Target Synonyms: Acetylneuraminyl hydrolase; exo-alpha-sialidase; G9 sialidase; Lysosomal sialidase; N acetyl alpha neuraminidase 1; N-acetyl-alpha-neuraminidase 1; NANH; NEU; NEU1; NEUR1_HUMAN; Neuraminidase 1; Neuraminidase; SIAL1; sialidase 1 (lysosomal sialidase); Sialidase 1; Sialidase; lysosomal; Sialidase-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8268