Learn More
Biomatik Corporation Human Sodium/potassium-transporting ATPase subunit alpha-1 (ATP1A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35586
Encompass_Preferred
Human Sodium/Potassium-Transporting Atpase Subunit Alpha-1 (Atp1A1) Elisa Kit for detecting Atpase, Na+/K+ Transporting, Alpha 1 Polypeptide in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05023; ATP1A1. Target Synonyms: A1A1; AT1A1; AT1A1_HUMAN; ATP1A1; Atpa-1; ATPase Na+/K+ transporting alpha 1 polypeptide; ATPase Na+/K+ transporting subunit alpha 1; BC010319; EC 3.6.3.9; MGC3285; MGC38419; MGC51750; Na K ATPase alpha A catalytic polypeptide; Na K ATPase catalytic subunit alpha A protein; Na (+)/K (+) ATPase 1; Na (+)/K (+) ATPase alpha-1 subunit; Na+; K+ ATPase alpha subunit; Na+/K+ ATPase alpha 1 subunit; Na+/K+ ATPase 1; Na; K ATPase alpha 1 subunit; Nkaa1b; Sodium potassium ATPase alpha 1 polypeptide; Sodium pump 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8271