Learn More
Biomatik Corporation Human soluble CD163 (sCD163) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35589
Encompass_Preferred
Human Soluble Cd163 (Scd163) Elisa Kit for detecting Cd163 Molecule in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86VB7; CD163. Target Synonyms: C163A_HUMAN; CD 163; CD163; CD163 antigen; CD163 molecule; Hemoglobin scavenger receptor; M130; M130 antigen precursor; Macrophage associated antigen; MM130; OTTHUMP00000238617; OTTHUMP00000238618; OTTHUMP00000238619; OTTHUMP00000238620; SCARI1; Scavenger receptor cysteine rich type 1 protein M130; sCD163; Soluble CD163. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8273