Learn More
Biomatik Corporation Human soluble OX40L (sOX40L) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35614
Encompass_Preferred
Human Soluble Ox40L (Sox40L) Elisa Kit for detecting Tumor Necrosis Factor (Ligand) Superfamily, Member 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P23510; TNFSF4. Target Synonyms: TNFSF4; TXGP1; Tumor necrosis factor ligand superfamily member 4; Glycoprotein Gp34; OX40 ligand; OX40L; TAX transcriptionally-activated glycoprotein 1; CD antigen CD252. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8280