Learn More
Biomatik Corporation Human soluble tumor necrosis factor-related apoptosis inducing ligand, sTRAIL ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35622
Encompass_Preferred
Human Soluble Tumor Necrosis Factor-Related Apoptosis Inducing Ligand, Strail Elisa Kit for detecting Tumor Necrosis Factor (Ligand) Superfamily, Member 10 in serum, plasma, cell culture supernates, urine, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P50591; TNFSF10. Target Synonyms: Apo 2 ligand; APO 2L; Apo-2 ligand; Apo-2L; APO2L; CD253; CD253 antigen; Chemokine tumor necrosis factor ligand superfamily member 10; Protein TRAIL; TL2; TNF Related Apoptosis Inducing Ligand; TNF related apoptosis inducing ligand TRAIL; TNF-related apoptosis-inducing ligand; TNF10_HUMAN; TNFSF10; TRAIL; Tumor necrosis factor (ligand) family member 10; Tumor Necrosis Factor (ligand) Superfamily Member 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8283