Learn More
Biomatik Corporation Human sonic hedgehog ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35630
Encompass_Preferred
Human Sonic Hedgehog Elisa Kit for detecting Sonic Hedgehog Homolog (Drosophila) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15465; SHH. Target Synonyms: HHG 1; HHG-1; HHG1; HLP 3; HLP3; Holoprosencephaly 3; HPE 3; HPE3; MCOPCB5; shh; SHH_HUMAN; SMMC I; SMMCI; Sonic Hedgehog (Drosophila) homolog; sonic hedgehog homolog (Drosophila); Sonic hedgehog homolog; Sonic hedgehog protein; Sonic hedgehog protein C-product; TPT; TPTPS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8285