Learn More
Biomatik Corporation Human Sortilin-related receptor (SORL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35636
Encompass_Preferred
Human Sortilin-Related Receptor (Sorl1) Elisa Kit for detecting Sortilin-Related Receptor(Sorl1) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92673; SORL1. Target Synonyms: C11orf32; FLJ21930; FLJ39258; gp250; LDLR relative with 11 ligand binding repeats; LDLR relative with 11 ligand-binding repeats; Low density lipoprotein receptor relative with 11 ligand binding repeats; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; LR 11; LR11; LRP 9; LRP9; Mosaic protein LR11; SORL 1; SORL_HUMAN; SORL1; SorLA; SorLA-1; Sortilin related receptor; Sortilin related receptor L (DLR class) A repeats containing; Sortilin-related receptor; Sorting protein related receptor containing LDLR class A repeats. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8287