Learn More
Biomatik Corporation Human SPARC (SPARC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35638
Encompass_Preferred
Human Sparc (Sparc) Elisa Kit for detecting Secreted Protein, Acidic, Cysteine-Rich (Osteonectin) in serum, plasma, tissue homogenates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09486; SPARC. Target Synonyms: AA517111; Basement membrane protein 40; Basement-membrane protein 40; BM 40; BM-40; BM40; Cysteine rich protein; hm:zeh0062; MGC128090; OI17; ON; Osteonectin; Secreted acidic cystein rich glycoprotein; Secreted protein acidic and cysteine rich; Secreted protein acidic and rich in cysteine; Secreted protein acidic cysteine rich (osteonectin); Secreted protein acidic cysteine rich; SPARC; SPRC; SPRC_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8288