Learn More
Biomatik Corporation Human SPARC-related modular calcium-binding protein 1 (SMOC1) ELISA Kit, 96 tests
Catalog No. 501498289
Encompass_Preferred
Human Sparc-Related Modular Calcium-Binding Protein 1 (Smoc1) Elisa Kit for detecting Sparc Related Modular Calcium Binding 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H4F8; SMOC1. Target Synonyms: Secreted modular calcium-binding protein 1; SMOC-1; SMOC1; SMOC1_HUMAN; SPARC-related modular calcium-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8289 Supplier Biomatik Corporation Supplier No. EKC35640
Add to Cart
Add to Cart
Human Sparc-Related Modular Calcium-Binding Protein 1 (Smoc1) Elisa Kit for detecting Sparc Related Modular Calcium Binding 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H4F8; SMOC1. Target Synonyms: Secreted modular calcium-binding protein 1; SMOC-1; SMOC1; SMOC1_HUMAN; SPARC-related modular calcium-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.