Learn More
Biomatik Corporation Human tartrate-resistant acid phosphatase 5b, TRACP-5b ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35711
Encompass_Preferred
Human Tartrate-Resistant Acid Phosphatase 5B, Tracp-5B Elisa Kit for detecting Acid Phosphatase 5, Tartrate Resistant in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20mIU/ml. Sensitivity: 0.078mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13686; ACP5. Target Synonyms: phosphatase; Acid phosphatase 5 tartrate resistant; Acid phosphatase 5; tartrate resistant; ACP5; EC 3.1.3.2; MGC117378; PPA5_HUMAN; serum band 5 tartrate-resistant acid phosphatase; SPENCDI; T5ap; Tartrate resistant acid ATPase; Tartrate resistant acid phosphatase type 5; Tartrate resistant acid phosphatase type 5 precursor; Tartrate-resistant acid ATPase; Tartrate-resistant acid phosphatase type 5; TR AP; TR-AP; TRACP 5; TRAP; TrATPase; Type 5 acid phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8298