Learn More
Biomatik Corporation Human Tau proteins ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35714
Encompass_Preferred
Human Tau Proteins Elisa Kit for detecting Microtubule-Associated Protein Tau in serum, plasma, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10636; MAPT. Target Synonyms: AI413597; AW045860; DDPAC; FLJ31424; FTDP 17; G protein beta1/gamma2 subunit interacting factor 1; MAPT; MAPTL; MGC134287; MGC138549; MGC156663; Microtubule associated protein tau; Microtubule associated protein tau isoform 4; Microtubule-associated protein tau; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; Paired helical filament tau; Paired helical filament-tau; PHF tau; PHF-tau; PPND; PPP1R103; Protein phosphatase 1; regulatory subunit 103; pTau; RNPTAU; TAU; TAU_HUMAN; Tauopathy and respiratory failure; Tauopathy and respiratory failure; included. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8299