Learn More
Biomatik Corporation Human T-cell surface protein tactile (CD96) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35729
Encompass_Preferred
Human T-Cell Surface Protein Tactile (Cd96) Elisa Kit for detecting Cd96 Molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40200; CD96. Target Synonyms: CD96; T-cell surface protein tactile; Cell surface antigen CD96; T cell-activated increased late expression protein; CD antigen CD96. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8300