Learn More
Biomatik Corporation Human Tenascin-X (TNXB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35740
Encompass_Preferred
Human Tenascin-X (Tnxb) Elisa Kit for detecting Tenascin Xb in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 46.88-3000ng/ml. Sensitivity: 11.7ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22105; TNXB. Target Synonyms: EDS3; Ehlers Danlos like syndrome; Growth inhibiting protein 45; Hexabrachion like protein; Hexabrachion-like protein; HXBL; NXB2; Tenascin X; Tenascin X precursor; Tenascin XB; Tenascin XB1; Tenascin XB2; Tenascin-X; TENX; TENX_HUMAN; TN X; TN-X; TNX; TNXB; TNXB1; TNXB2; TNXBS; VUR8; XB; XBS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8302