Learn More
Biomatik Corporation Human terminal complement complex SC5b9 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35746
Encompass_Preferred
Human Terminal Complement Complex Sc5B9 Elisa Kit for detecting Terminal Complement Complex Sc5B9 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000ng/ml. Sensitivity: 7.81ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8304