Learn More
Biomatik Corporation Human thiopurine S-methyltransferase (TPMT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35756
Encompass_Preferred
Human Thiopurine S-Methyltransferase (Tpmt) Elisa Kit for detecting Thiopurine S-Methyltransferase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100mU/ml. Sensitivity: 0.39mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51580; TPMT. Target Synonyms: HGNC:12014; S adenosyl L methionine thiopurine S methyltransferase; Thiopurine methyltransferase; Thiopurine S methyltransferase; Thiopurine S-methyltransferase; TPMT; TPMT_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8305