Learn More
Biomatik Corporation Human thymic stromal lymphopoietin, TSLP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35777
Encompass_Preferred
Human Thymic Stromal Lymphopoietin, Tslp Elisa Kit for detecting Thymic Stromal Lymphopoietin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q969D9; TSLP. Target Synonyms: Thymic stromal lymphopoietin; Thymic stromal lymphopoietin protein TSLP; Tslp; TSLP protein; TSLP_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8313