Learn More
Biomatik Corporation Human tissue inhibitors of metalloproteinase 2, TIMP-2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35797
Encompass_Preferred
Human Tissue Inhibitors Of Metalloproteinase 2, Timp-2 Elisa Kit for detecting Timp Metallopeptidase Inhibitor 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16035; TIMP2. Target Synonyms: CSC 21K; CSC-21K; CSC21K; Metalloproteinase inhibitor 2; Metalloproteinase inhibitor 2 precursor; TIMP 2; TIMP metallopeptidase inhibitor 2; TIMP-2; TIMP2; TIMP2_HUMAN; Tissue Inhibitor of Metalloproteinase 2; Tissue inhibitor of metalloproteinases 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8317