Learn More
Biomatik Corporation Human Tissue-type Plasminogen Actilyse, t-PA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35800
Encompass_Preferred
Human Tissue-Type Plasiminogen Actilyse, T-Pa Elisa Kit for detecting Plasminogen Activator, Tissue in serum, palsma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00750; PLAT. Target Synonyms: Alteplase; DKFZp686I03148; Plasminogen activator tissue; Plasminogen activator tissue type; PLAT; Reteplase; t PA; T Plasminogen Activator; t-PA; T-plasminogen activator; Tissue plasminogen activator (t PA); Tissue type plasminogen activator; Tissue-type plasminogen activator chain B; tPA; TPA_HUMAN; TPA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8318