Learn More
Biomatik Corporation Human transforming growth factors 2, TGF2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35847
Encompass_Preferred
Human Transforming Growth Factors Β2, Tgfβ2 Elisa Kit for detecting Transforming Growth Factor, Beta 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P61812; TGFB2. Target Synonyms: MGC116892; TGF-beta2; transforming growth factor-beta2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8321