Learn More
Biomatik Corporation Human transglutaminase 2C polypeptide, TGM2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35851
Encompass_Preferred
Human Transglutaminase 2C Polypeptide, Tgm2 Elisa Kit for detecting Transglutaminase 2 (C Polypeptide, Protein-Glutamine-Gamma-Glutamyltransferase) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.68pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21980; TGM2. Target Synonyms: ALPHA SUBUNIT; C polypeptide; EC 2.3.2.13; epididymis secretory protein Li 45; G alpha h; G[a]h; Gh CLASS G ALPHA h; GNAH; GNAH G PROTEIN; H POLYPEPTIDE; HEL-S-45; Protein glutamine gamma glutamyltransferase 2; Protein-glutamine gamma-glutamyltransferase 2; TG 2; TG (C); TG2; TGase C; TGase H; TGase-2; TgaseII; TGC; TGC GUANINE NUCLEOTIDE BINDING PROTEIN; TGM2; TGM2_HUMAN; Tissue transglutaminase; Transglutaminase 2; Transglutaminase 2 C polypeptide; Transglutaminase C; Transglutaminase H; Transglutaminase-2; tTG; tTGas. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8322