Learn More
Biomatik Corporation Human Translationally-controlled tumor protein (TPT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35862
Encompass_Preferred
Human Translationally-Controlled Tumor Protein (Tpt1) Elisa Kit for detecting Tumor Protein, Translationally-Controlled 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13693; TPT1. Target Synonyms: FLJ27337; Fortilin; Histamine releasing factor; Histamine-releasing factor; HRF; Immunoglobulin E-dependent; p02; p23; TCTP; TCTP_HUMAN; tpt1; Translationally controlled tumor protein; Translationally-controlled tumor protein; Tumor protein translationally controlled 1; tumor protein; translationally-controlled 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8323