Learn More
Biomatik Corporation Human transthyretin (TTR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35871
Encompass_Preferred
Human Transthyretin (Ttr) Elisa Kit for detecting Transthyretin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-25ug/ml. Sensitivity: 0.78ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02766; TTR. Target Synonyms: Amyloid polyneuropathy; Amyloidosis I; ATTR; Carpal tunnel syndrome 1; CTS; CTS1; Dysprealbuminemic euthyroidal hyperthyroxinemia; Dystransthyretinemic hyperthyroxinemia; Epididymis luminal protein 111; HEL111; HsT2651; PALB; Prealbumin amyloidosis type I; Prealbumin; Prealbumin Thyroxine-binding; Senile systemic amyloidosis; TBPA; Thyroxine binding prealbumin; Transthyretin; TTHY_HUMAN; TTR; TTR protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8325