Learn More
Biomatik Corporation Human Tripartite motif-containing protein 72 (TRIM72) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35890
Encompass_Preferred
Human Tripartite Motif-Containing Protein 72 (Trim72) Elisa Kit for detecting Tripartite Motif-Containing 72 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6ZMU5; TRIM72. Target Synonyms: LOC493829; Mg53; Mitsugumin-53; TRI72_HUMAN; trim72; Tripartite motif containing 72; Tripartite motif containing protein 72; Tripartite motif-containing protein 72. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8327