Learn More
Biomatik Corporation Human Tropomyosin alpha-4 chain (TPM4) ELISA Kit, 96 tests
Catalog No. 501498330
Encompass_Preferred
Human Tropomyosin Alpha-4 Chain (Tpm4) Elisa Kit for detecting Tropomyosin 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P67936; TPM4. Target Synonyms: epididymis secretory protein Li 108; HEL-S-108; TM30p1; TMP 4; TPM4; TPM4_HUMAN; Tropomyosin 4; Tropomyosin alpha 4 chain; Tropomyosin alpha-4 chain; Tropomyosin-4; wu:fb06e07; zgc:103436; zgc:63909. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8330 Supplier Biomatik Corporation Supplier No. EKC35896
Human Tropomyosin Alpha-4 Chain (Tpm4) Elisa Kit for detecting Tropomyosin 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P67936; TPM4. Target Synonyms: epididymis secretory protein Li 108; HEL-S-108; TM30p1; TMP 4; TPM4; TPM4_HUMAN; Tropomyosin 4; Tropomyosin alpha 4 chain; Tropomyosin alpha-4 chain; Tropomyosin-4; wu:fb06e07; zgc:103436; zgc:63909. Restrictions: For Research Use Only. Not for use in diagnostic procedures.