Learn More
Biomatik Corporation Human trypsin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35902
Encompass_Preferred
Human Trypsin Elisa Kit for detecting Protease, Serine, 1 (Trypsin 1) in serum, plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07477; PRSS1. Target Synonyms: Alpha-trypsin chain 2; Beta-trypsin; Cationic trypsinogen; Digestive zymogen; Nonfunctional trypsin 1; Prss1; Serine protease 1; TCR V beta 4.1; TRP1; TRY1; TRY1_HUMAN; TRY4; TRYP1; Trypsin I; Trypsinogen 1; Trypsinogen A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8331