Learn More
Biomatik Corporation Human tryptophanyl-tRNA synthetase, WARS ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35910
Encompass_Preferred
Human Tryptophanyl-Trna Synthetase, Wars Elisa Kit for detecting Tryptophanyl-Trna Synthetase in serum, plasma, cell lysates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P23381; WARS. Target Synonyms: Gamma 2; GAMMA-2; Gamma2; hWRS; IFI 53; IFI53; IFP 53; IFP53; Interferon induced protein 53; Interferon-induced protein 53; SYWC_HUMAN; T2-TrpRS; TrpRS; Tryptophan tRNA ligase; cytoplasmic; Tryptophan tRNA ligase 1 cytoplasmic; Tryptophan tRNA ligase; Tryptophan--tRNA ligase; Tryptophanyl tRNA synthetase; Tryptophanyl tRNA synthetase cytoplasmic; WARS; WARS protein; WRS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8332