Learn More
Biomatik Corporation Human Tumor necrosis factor receptor superfamily member 6B (TNFRSF6B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35928
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily Member 6B (Tnfrsf6B) Elisa Kit for detecting Tumor Necrosis Factor Receptor Superfamily, Member 6B, Decoy in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95407; TNFRSF6B. Target Synonyms: DcR3; Decoy receptor 3; Decoy receptor for Fas ligand; DJ583P15.1.1; M68; M68E; OTTHUMP00000031583; TNF6B_HUMAN; TNFRSF6B; TR6; Tumor necrosis factor receptor superfamily member 6B; Tumor necrosis factor receptor superfamily member 6b decoy; UNQ186/PRO212. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8334