Learn More
Biomatik Corporation Human tumor necrosis factor superfamily 15 (TL1A/TNFSF15) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35930
Encompass_Preferred
Human Tumor Necrosis Factor Superfamily 15 (Tl1A/Tnfsf15) Elisa Kit for detecting Tumor Necrosis Factor (Ligand) Superfamily, Member 15 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95150; TNFSF15. Target Synonyms: TNFSF15; TL1; VEGI; Tumor necrosis factor ligand superfamily member 15; TNF ligand-related molecule 1; Vascular endothelial cell growth inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8335