Learn More
Biomatik Corporation Human tumor necrosis factor-associated factor 1, TRAF1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35934
Encompass_Preferred
Human Tumor Necrosis Factor-Associated Factor 1, Traf1 Elisa Kit for detecting Tnf Receptor-Associated Factor 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13077; TRAF1. Target Synonyms: 4732496E14Rik; EBI6; EBV induced protein 6; Epstein-Barr virus-induced protein 6; MGC:10353; Tnf receptor associated factor 1; TNF receptor-associated factor 1; TRAF 1; TRAF1; TRAF1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8336