Learn More
Biomatik Corporation Human Tumor Necrosis Factor-Related Apoptosis-Inducing Ligand Receptor 2 (TRAIL-R2/DR5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35937
Encompass_Preferred
Human Tumor Necrosis Factor-Related Apoptosis-Inducing Ligand Receptor 2 (Trail-R2/Dr5) Elisa Kit for detecting Tumor Necrosis Factor Receptor Superfamily, Member 10B in serum, plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O14763; TNFRSF10B. Target Synonyms: TNFRSF10B; DR5; KILLER; TRAILR2; TRICK2; ZTNFR9; UNQ160/PRO186; Tumor necrosis factor receptor superfamily member 10B; Death receptor 5; TNF-related apoptosis-inducing ligand receptor 2; TRAIL receptor 2; TRAIL-R2; CD antigen CD262. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8339