Learn More
Biomatik Corporation Human type collagen helical peptide, HELIX- ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35946
Encompass_Preferred
Human Type Ii Collagen Helical Peptide, Helix-Ii Elisa Kit for detecting Type II Collagen Helical Peptide, Helix-II in serum, plasma, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.312ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: type 2 collagen helical peptide; HELIX-2; type II collagen helical peptide; HELIX-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8342